DL-Asparagine monohydrate 99,0 Titrimetric

Supplier: TCI America

Synonyms: DL-Aspartic acid 4-amide monohydrate, H-DL-Asn-OH monohydrate, (±)-2-Aminosuccinic acid 4-amide monohydrate, DL-Asparagine hydrate, H-DL-Asn-OH·H₂O, DL-2-Aminosuccinamic acid monohydrate, 2,4-Diamino-4-oxobutanoic acid monohydrate

A0541-25G A0541-500G A0541-100G
TCA0541-025GEA 40.53 CAD
TCA0541-025G TCA0541-500G TCA0541-100G
DL-Asparagine monohydrate 99,0 Titrimetric
DL-Asparagine monohydrate

CAS Number: 3130-87-8
MDL Number: MFCD00151039
Molecular Formula: C4H8N2O3
Molecular Weight: 132.12
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Asn-OH

Formula: C₄H₈N₂O₃
MW: 150.13 g/mol
MDL Number: MFCD00151039
CAS Number: 3130-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78d05d0cff54fa28646eabedd79483c9aa0b0640&vid=575a2241581d39d64fb2adec186801a0f3bedc9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR