D-(-)-Isoascorbic acid 98,0

Supplier: TCI America

Synonyms: Erythorbic acid, (R)-5-((R)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, D-Isoascorbic acid

A0520-25G A0520-500G
TCA0520-025GEA 28.88 CAD
TCA0520-025G TCA0520-500G
D-(-)-Isoascorbic acid 98,0
D(-)-Isoascorbic acid

CAS Number: 89-65-6
MDL Number: MFCD00005378
Molecular Formula: C6H8O6
Molecular Weight: 176.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 167
Specific rotation [a]20/D: -17.5 deg (C=10, H2O)


Product Categories: Bioscience, Sugar Acids, Sugars (Others), Vitamin Derivatives
Other Product Names / Synonyms: D-Erythorbic Acid, D-Isoascorbic Acid

Formula: C₆H₈O₆
MW: 176.12 g/mol
Melting Pt: 167 °C
MDL Number: MFCD00005378
CAS Number: 89-65-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbd57834ee0719ce2bff91af8c97a465335486d9&vid=dbd57834ee0719ce2bff91af8c97a465335486d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR