L(-)-Arabitol ≥97.0%
Supplier: TCI America
Synonyms:
L-(-)-Arabinitol, (2S,4S)-Pentane-1,2,3,4,5-pentaol, L-(-)-Arabitol, L-Arabinitol
A0518-1G
A0518-25G
A0518-5G
TCA0518-001GEA
45.82
CAD
TCA0518-001G
TCA0518-025G
TCA0518-5G
L(-)-Arabitol ≥97.0%
L(-)-Arabitol
CAS Number: 7643-75-6
MDL Number: MFCD00064290
Molecular Formula: C5H12O5
Molecular Weight: 152.15
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 103
Specific rotation [a]20/D: -12 deg (C=5, Sodium borate(8%))
Product Categories: Bioscience, Sugar Alcohols, Arabinoses
Other Product Names / Synonyms: L-(-)-Arabinitol
Formula:
C₅H₁₂O₅ Melting Pt: 103 °C |
MDL Number:
MFCD00064290 CAS Number: 7643-75-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...