D-(+)-Arabitol ≥98.0%
Supplier: TCI America
Synonyms:
D-Arabitol, (2R,4R)-Pentane-1,2,3,4,5-pentaol
A0516-1G
A0516-25G
TCA0516-001GEA
41.08
CAD
TCA0516-001G
TCA0516-025G
D-(+)-Arabitol ≥98.0%
D(+)-Arabitol
CAS Number: 488-82-4
MDL Number: MFCD00004709
Molecular Formula: C5H12O5
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 103
Specific rotation [a]20/D: 12 deg (C=5, Sodium borate(8%))
Product Categories: Bioscience, Sugar Alcohols, Arabinoses
Other Product Names / Synonyms: D-(+)-Arabinitol
Formula:
C₅H₁₂O₅ MW: 152.15 g/mol Melting Pt: 103 °C |
MDL Number:
MFCD00004709 CAS Number: 488-82-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...