D-(-)-Arabinose ≥99.0%

Supplier: TCI America

Synonyms: D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal

A0513-25G A0513-100G
TCA0513-025GEA 54.87 CAD
TCA0513-025G TCA0513-100G
D-(-)-Arabinose ≥99.0%
D(-)-Arabinose

CAS Number: 10323-20-3
MDL Number: MFCD00135608
Molecular Formula: C5H10O5
Molecular Weight: 150.13
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 161
Specific rotation [a]20/D: -104 deg (C=10, H2O)

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Melting Pt: 161 °C
MDL Number: MFCD00135608
CAS Number: 10323-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c50d784a26c928b88ef23dfd533c830e8b08c54d&vid=c50d784a26c928b88ef23dfd533c830e8b08c54d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR