Sodium Anthraquinone-2-sulfonate Monohydrate ≥98.0%
Supplier: TCI America
Synonyms:
Anthraquinone-2-sulphonic acid sodium salt monohydrate, Anthraquinone-2-sulfonic acid sodium salt monohydrate
A0508-25G
A0508-500G
TCA0508-025GEA
61.49
CAD
TCA0508-025G
TCA0508-500G
Sodium Anthraquinone-2-sulfonate Monohydrate ≥98.0%
Sodium Anthraquinone-2-sulfonate Monohydrate
CAS Number: 153277-35-1
MDL Number: MFCD00149068
Molecular Formula: C14H8O5S
Molecular Weight: 310.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: Anthraquinone-2-sulfonic Acid Sodium Salt
Formula:
C₁₄H₈O₅S |
MDL Number:
MFCD00149068 CAS Number: 153277-35-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...