4-Hydroxydiphenylamine ≥98.0%
Supplier: TCI America
Synonyms:
4-(Phenylamino)phenol
A0470-500G
A0470-25G
TCA0470-500GEA
1131.28
CAD
TCA0470-500G
TCA0470-025G
4-Hydroxydiphenylamine ≥98.0%
4-Hydroxydiphenylamine
CAS Number: 122-37-2
MDL Number: MFCD00020142
Molecular Formula: C12H11NO
Molecular Weight: 185.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 165
Melting point (°C): 73
Flash Point (°C): 209
Other Product Names / Synonyms: p-Anilinophenol
Formula:
C₁₂H₁₁NO MW: 185.23 g/mol Boiling Pt: 165 °C Melting Pt: 73 °C Flash Pt: 209 °C |
MDL Number:
MFCD00020142 CAS Number: 122-37-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...