2-Amino-1-naphthalenesulfonic acid ≥98.0%
Supplier: TCI America
Synonyms:
2-Aminonaphthalene-1-sulphonic acid, 2-Aminonaphthalene-1-sulfonic acid
A0343-25G
A0343-500G
TCA0343-025GEA
77.17
CAD
TCA0343-025G
TCA0343-500G
2-Amino-1-naphthalenesulfonic acid ≥98.0%
2-Amino-1-naphthalenesulphonic acid
CAS Number: 81-16-3
MDL Number: MFCD00003988
Molecular Formula: C10H9NO3S
Molecular Weight: 223.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 2-Naphthylamine-1-sulfonic Acid, Tobias Acid
Formula:
C₁₀H₉NO₃S |
MDL Number:
MFCD00003988 CAS Number: 81-16-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...