2-Amino-5-methylbenzenesulfonic acid ≥98.0%
Supplier: TCI America
Synonyms:
4-Aminotoluene-3-sulfonic acid, 4-Aminotoluene-3-sulphonic acid
A0328-500G
A0328-25G
TCA0328-500GEA
135.88
CAD
TCA0328-500G
TCA0328-25G
2-Amino-5-methylbenzenesulfonic acid ≥98.0%
2-Amino-5-methylbenzenesulphonic acid
CAS Number: 88-44-8
MDL Number: MFCD00007908
Molecular Formula: C7H9NO3S
Molecular Weight: 187.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 4-Aminotoluene-3-sulfonic Acid, 4-Methylaniline-2-sulfonic Acid, 2-Amino-5-methylbenzenesulfonic Acid
Formula:
C₇H₉NO₃S |
MDL Number:
MFCD00007908 CAS Number: 88-44-8 UN: 3261 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...