7-Aminoenanthic acid ≥98.0%

Supplier: TCI America

Synonyms: 7-Aminoheptanoic acid, 7-Aminoheptylic acid

A0311-1G A0311-5G A0311-25G
TCA0311-1GEA 178.54 CAD
TCA0311-1G TCA0311-5G TCA0311-25G
7-Aminoenanthic acid ≥98.0%
7-Aminoenanthic acid

CAS Number: 929-17-9
MDL Number: MFCD00008242
Molecular Formula: C7H15NO2
Molecular Weight: 145.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 189


Other Product Names / Synonyms: 7-Aminoenanthic Acid

Formula: C₇H₁₅NO₂
MW: 145.2
Melting Pt: 189 °C
MDL Number: MFCD00008242
CAS Number: 929-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7251efc1702f49787ef1700d29da0e97f29bcb0&vid=a7251efc1702f49787ef1700d29da0e97f29bcb0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR