N-Acetyl-L-leucine ≥99.0%

Supplier: TCI America

Synonyms: N-Ac-Leu-OH, Ac-Leu-OH

A0098-5G A0098-25G
TCA0098-5GEA 64.78 CAD
TCA0098-5G TCA0098-025G
N-Acetyl-L-leucine ≥99.0%
N-Acetyl-L-leucine

CAS Number: 1188-21-2
MDL Number: MFCD00065131
Molecular Formula: C8H15NO3
Molecular Weight: 173.21
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 185
Specific rotation [a]20/D: -22 deg (C=5, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-Leu-OH

Formula: C₈H₁₅NO₃
MW: 173.2 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00065131
CAS Number: 1188-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a96a0bcc26835fc75dc3ddb362702b838c4badd1&vid=a96a0bcc26835fc75dc3ddb362702b838c4badd1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR