N-Acetyl-DL-leucine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

A0097-25G A0097-5G
TCA0097-025GEA 93.22 CAD
TCA0097-025G TCA0097-5G
N-Acetyl-DL-leucine ≥99.0% (by titrimetric analysis)
N-Acetyl-DL-leucine

CAS Number: 99-15-0
MDL Number: MFCD00026498
Molecular Formula: C8H15NO3
Molecular Weight: 173.21
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 160


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-DL-Leu-OH

Formula: C₈H₁₅NO₃
Melting Pt: 160 °C
MDL Number: MFCD00026498
CAS Number: 99-15-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62d5e9d3a4624db77fad5be684488de6e39c7d7f&vid=62d5e9d3a4624db77fad5be684488de6e39c7d7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR