N-Acetyl-ε-caprolactam ≥98.0%
Supplier: TCI America
A0081-500G
A0081-25G
TCA0081-500GEA
379.2
CAD
TCA0081-500G
TCA0081-025G
N-Acetyl-ε-caprolactam ≥98.0%
N-Acetyl-epsilon-caprolactam
CAS Number: 1888-91-1
MDL Number: MFCD00003262
Molecular Formula: C8H13NO2
Molecular Weight: 155.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 113
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: N-Acetyl-2-oxohexamethyleneimine
Formula:
C₈H₁₃NO₂ Boiling Pt: 134 °C Density: 1.10 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00003262 CAS Number: 1888-91-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...