2-Acetamidoethanol ≥95.0%

Supplier: TCI America

A0075-25G A0075-500G
TCA0075-025GEA 53.72 CAD
TCA0075-025G TCA0075-500G
2-Acetamidoethanol ≥95.0%
2-Acetamidoethanol

CAS Number: 142-26-7
MDL Number: MFCD00002836
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 167
Flash Point (°C): 177
Freezing point (°C): 16
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: N-Acetylethanolamine, N-(2-Hydroxyethyl)acetamide

Formula: C₄H₉NO₂
Boiling Pt: 167 °C
Density: 1.12 (20 °C)
Flash Pt: 177 °C
MDL Number: MFCD00002836
CAS Number: 142-26-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a62d87e271aee5e7189f4337a88ea34c7bda6fb&vid=4a62d87e271aee5e7189f4337a88ea34c7bda6fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR