Acetone semicarbazone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0058-500G A0058-25G
TCA0058-500GEA 374.46 CAD
TCA0058-500G TCA0058-025G
Acetone semicarbazone ≥98.0% (by titrimetric analysis)
Acetone semicarbazone

CAS Number: 110-20-3
MDL Number: MFCD00014785
Molecular Formula: C4H9N3O
Molecular Weight: 115.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 187

Formula: C₄H₉N₃O
Melting Pt: 187 °C
MDL Number: MFCD00014785
CAS Number: 110-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0120c3edefc13c099457e2cec6a84baf5df817d5&vid=0120c3edefc13c099457e2cec6a84baf5df817d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR