Acetohydroxamic acid ≥98.0%

Supplier: TCI America

Synonyms: N-Hydroxyacetamide

A0051-25G A0051-10G
TCA0051-25GEA 213.3 CAD
TCA0051-25G TCA0051-010G
Acetohydroxamic acid ≥98.0%
Acetohydroxamic acid

CAS Number: 546-88-3
MDL Number: MFCD00009994
Molecular Formula: C2H5NO2
Molecular Weight: 75.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 92


Other Product Names / Synonyms: N-Hydroxyacetamide

Formula: C₂H₅NO₂
MW: 75.0666 g/mol
Melting Pt: 92 °C
MDL Number: MFCD00009994
CAS Number: 546-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eeecd0eaecef85da56b7040c3bd71486c620f290&vid=eeecd0eaecef85da56b7040c3bd71486c620f290&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR