GL Screwthread Connectors, Ace Glass Incorporated
Supplier: Ace Glass
7620-14 7620-18 7620-25 7620-32 7620-45 7620-60
89059-762EA 21.18 CAD
89059-762 89059-764 89059-766 89059-768 89059-770 89059-772
GL Screwthread Connectors, Ace Glass Incorporated
Fittings and Connectors
Externally threaded glass connectors can be used as replacements on apparatus using the GL thread, or when designing items where an external thread is preferred.