The 5 Gallon Multiple Polypropylene Bucket System features a compact electropolished stainless steel rolling cart with two autoclavable polypropylene buckets. A third bucket is available for use when a three bucket system is required.
- Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
- Compatible with all Contec mops, depending upon wringer choice
- System can be used with two or three buckets
- Handle can be removed for storage
- Compatible with most disinfectants
Compact design improves maneuverability and reduces storage space. The durable yet lightweight cart has four casters, two that lock easily with the touch of a foot. An extendable frame pulls out from the front of the cart to accommodate the third bucket. Includes a stainless steel connecting kit to prevent drips between the buckets.
The cart features a unique removable handle for easier autoclaving and storage. For maximum flexibility, we offer two types of wringers that are ordered separately. Choose from a stainless steel downpress wringer, or a stainless steel wringer that fits directly on the waste bucket.
Ordering information: Wringer options must be ordered separately. See the Wringers, Contec® product page for compatible options.