ProSieve™ Protein Markers
Supplier: Lonza
ProSieve™
00193837 00193837
89125-526EA 697.56 CAD
89125-526 CA89125-526
ProSieve™ Protein Markers
Electrophoresis Reagents Molecular Weight Markers
The ProSieve® QuadColor® Protein marker is a mixture of 12 recombinant, highly purified proteins with molecular weights of 4, 6, 10, 15, 25, 40, 55, 70, 100, 140, 170, 250, and 300 kDa.
  • Ready-to- use - no heating, further dilution or addition of a reducing agent is required before use
  • Brightly colored ladder - easy to rember pattern
ProSieve™ QuadColor™ Protein Marker, just add water and load for visible protein confirmation based on the multi-colored identification band patterns.
12 month shelf life and freezer storage.
Sustainable and considered non toxic, but should be disposed with proper safety guidelines.