Sterile TechniSat® 70% IPA Saturated Wipers
Supplier: Texwipe
TX3214 TX3217 TX3217
CA-TWTX3217CS 4120.04 CAD
CA-TWTX3217 T-WTX3214 TWTX3217
Sterile TechniSat® 70% IPA Saturated Wipers
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
These convenient wipers offer portion control and reduced potential of spillage in and out of the cleanroom.
  • Fibers blended by a hydroentanglement procedure that uses water instead of chemicals to bind
  • Excellent bidirectional strength and a very low extractable level
Sterile TechniSat® is used for alcohol wiping in sterile environments in pharmaceutical aseptic fill areas, biotechnology manufacturing facilities, microbiological laboratories, sterile suites and prep rooms.
Sterile wipers are saturated with 70% USP grade isopropyl alcohol and packaged in resealable flexpacks. The flexpacks are then double-bagged to maintain sterility. Each package is gamma irradiated to 10 to 6 sterility assurance level. Wipers are lot controlled for traceability.
The blended material of 55% cellulose and 45% polyester provides the high absorbency of a natural fiber with the cleanliness and strength of a synthetic material.