Head, Hickman Still, Ace Glass
Supplier: Ace Glass
9576-02 9576-04
89075-186EA 181.25 CAD
89075-186 89075-188
Head, Hickman Still, Ace Glass
Distillation Components Distillation Heads
This microscale Hickman still head is used to carry out micro-sample fractional distillations
The top joint is a combination cap, thread and standard taper joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external thread to facilitate an o-ring, and an open-top phenolic cap. The internal (or I.D.) at the top is a ground outer joint, at bottom is an inner [ST] joint. The inner joint can be secured in reaction vials via “O-Ring-CAP-SEAL” connection. Fabricated from borosilicate glass.
Ordering information: Supplied complete with cap, o-ring, andMore Product Information