Laboratory Support, Ace Glass
Supplier: Ace Glass
11214-20
89075-728EA 857.7 CAD
89075-728
Laboratory Support, Ace Glass
Laboratory Jacks
Fabricated from corrosion- and chemical-resistant, glass-reinforced polypropylene and PTFE filled polyethermide, this lightweight support is non-magnetic and non-conducting
It utilizes the scissors jack principle for quick, easy and accurate height adjustment. The PTFE nuts provide smooth, non-freezing operation. The jack can be adjusted in height.