Corning® PureCoat™ ECM Mimetic Flasks, Fibronectin Peptide, Corning
Supplier: Corning
356242 356243
CA89237-474CS 612.47 CAD
CA89237-474 CA89237-476
Corning® PureCoat™ ECM Mimetic Flasks, Fibronectin Peptide, Corning
Flasks Cell Culture Flasks
Corning® PureCoat™ ECM Mimetic Flasks, Fibronectin Peptide, Corning® PureCoat™ ECM Mimetic Cultureware is coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent alternative to complex extracellular peptides.
  • Animal-free, pre-coated, synthetic peptide surface for predictable scale-up
  • Streamlines cell culture workflow, reducing handling and contamination risk
  • Even distribution of media across all layers for homogeneous cell growth
  • Ability to mix cells and reagents in the Multi-Flask saves time and reduces risk of contamination
  • Flexible design lets you pour or aspirate/recover cells using a pipet
Certifications: Manufactured in compliance to cGMP and ISO 9001:2008, ISO 13485:2012 standards.