VWR® Battery Hydrometer with Siphon Set, Traceable to NIST
Supplier: VWR
61809-0211
34605-120EA 232.98 CAD
34605-120
VWR® Battery Hydrometer with Siphon Set, Traceable to NIST
Hydrometers
Used to determine the specific gravity / relative density (g/cmᶟ) of battery acid, this hydrometer comes with a siphon set to collect battery acid for an accurate reading.
  • Rubber parts are made of nitrile, a synthetic rubber that is more chemical resistant and more durable than conventional rubber
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 77°F
  • Hand inspected by our U.S. quality team
Made in the USA, and packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Certifications: Traceable to NIST.
Caution: Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.