FoamZorb™ Foam Wipes, Contec
Supplier: Contec
FZ451/8
33500-504CS 1036.7 CAD
33500-504
FoamZorb™ Foam Wipes, Contec
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Hydrophilic polyurethane foam, high sorptive capacity with low levels of extractable residues and particles. For spill cleanup or solution/solvent application.
Packaging: 18/bag, 56 Bags/case.