VWR® Pocket Liquid-In-Glass Thermometers
Supplier: VWR International
CA89095-704 CA89095-708
CA89095-704CS 1365.93 CAD
CA89095-704 CA89095-708
VWR® Pocket Liquid-In-Glass Thermometers
Thermometers Analog Thermometers
Featuring a closed metal case to protect from breakage, these total immersion thermometers are a compact 160mm (6.3”), making them ideal for field studies, waste water treatment and any “on-the-go” applications
Mercury free
Contain blue organic liquid against lead-free, white back glass
Tested and inspected by our USA quality team
Certifications: Traceable to NIST.
Ordering information: Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Order Now
SPECIFICATIONS
Environmentally Preferable