Chromatography Column with Fritted Disc, #15 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5905-25 5905-41 5905-39 5905-29 5905-15 5905-46 5905-49 5905-35 5905-19 5905-23
89053-510EA 700.66 CAD
89053-510 89053-522 89053-538 89053-542 89053-566 89053-572 89053-574 89053-576 89053-578 89053-580
Chromatography Column with Fritted Disc, #15 Ace-Thred, Ace Glass Incorporated
Chromatography Columns
This chromatography column has a 2 mm bore, 1:5 taper solid PTFE stopcock plug, a Porosity B (70 to 100 µm) fritted disc sealed in at bottom, and #15 Ace-Thred at the top of column.
The column is supplied with a nylon bushing (89059-220) and FETFE O-Ring. Fabricated from borosilicate glass.