Chromatography Column, 1:PTFE Plug, #15 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5908-06 5908-12 5908-18 5908-22 5908-36 5908-42 5908-44 5908-46
89053-634EA 460.6 CAD
89053-634 89053-636 89053-638 89053-640 89053-642 89053-644 89053-646 89053-648
Chromatography Column, 1:PTFE Plug, #15 Ace-Thred, Ace Glass Incorporated
Chromatography Columns
This chromatography column has an integral reservoir, a #15 Ace-Thred connection at top, and a 2 mm bore, 1:5 taper PTFE plug at bottom.
The column is supplied with a nylon bushing (89059-220) and FETFE O-Ring. Fabricated from borosilicate glass.
Ordering information: Complete item consists of column with nylon bushing (89059-220) and O-Ring, and tubing connector (89052-938) for connecting 12.5 to 14 mm ext.Ø tubing to column. Or you can purchase column with bushing and O-Ring only.