Three-Way Tubing Connector Valve, Ace Glass Incorporated
Supplier: Ace Glass
12730-39
89077-462EA 873.07 CAD
89077-462
Three-Way Tubing Connector Valve, Ace Glass Incorporated
Valves Non Return Valves
This three-way tubing connector valve is used for applications where stream switching, reagent switching, sample removal, or sample injection is desirable
The body is machined from a single piece of PTFE. The Tefzel valves are rated to 500psi, and they have one moving part per channel to ensure trouble-free operation and a long service life. The 1.5mm I.D. channels meet at valve center. Leak-tight to outside even at excessive pressures. Dead space within the valve is zero. Caps are polypropylene with Fluoron elastomer O-Ring seals for instant connection of rigid or flexible tubing within a range of 0.5 to 4mm O.D. Equipped with ¼"–28 UNF thread at the back of the caps for direct connection with other adapters. This valve is autoclavable.