Trubore® Bearing with Hose Connection, Ace-Thread, Ace Glass Incorporated
Supplier: Ace Glass
8066-70 8066-71 8066-72 8066-79 8066-73 8066-130 8066-132 8066-134 8066-140 8066-136
89082-486EA 141.01 CAD
89082-486 89082-488 89082-490 89082-492 89082-494 89082-918 89082-920 89082-922 89082-924 89082-926
Trubore® Bearing with Hose Connection, Ace-Thread, Ace Glass Incorporated
Stirrers Stirring Tools
This self-aligning, lubricant-free PTFE Trubore® bearing is designed for use with stainless steel or glass 10 mm stirring shafts
  • The lock nut permits stirring in either direction without fear of unthreading
  • The compression saddle with O-Ring maintains constant force with little attention
  • Accepts vacuum down to 1 mm
  • Not recommended for use with precision ground glass shafts
  • Fabricated from borosilicate glass
An added side port hose connection enables the purging of flask contents with a gas, or allows for an inert gas blanket in airless work.