Trubore® Bearing, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
8066-32 8066-06 8066-20 8066-24 8066-28 8066-33 8066-43 8066-46 8066-50 8066-55 8066-60
89061-848EA 185.32 CAD
89061-848 89061-854 89061-872 89061-874 89061-876 89061-880 89061-882 89061-884 89061-886 89097-170 89097-172
Trubore® Bearing, PTFE, Ace Glass Incorporated
Stirrers Stirring Tools
This self-aligning, lubricant-free PTFE Trubore® glass and PTFE bearing is designed for use with stainless steel or glass 10 mm stirring shafts
  • The compression saddle with O-Ring maintains constant force with little attention
  • Vacuum down to 1 mm
  • Not recommended for precision ground glass shafts
  • Fabricated from borosilicate glass
The lock nut permits stirring in either direction without fear of unthreading.
Delivery information: Available as a complete item, or individual components may be purchased separately. The complete item includes threaded glass body/adapter, 89061-854 PTFE inner bearing, 89061-864 glass-filled PTFE bushing, 89061-868 glass-filled PTFE lock nut, and 89061-870 Rulon compression saddle with O-Ring. Use with glass stirring shafts such as 89061-960 or 89061-970 (among others). For replacement compression saddle size -111 FETFE O-Rings, see 89060-988 series. For size -014 bearing replacement FETFE O-Ring, see 89060-976 series.