BRAND BRANDplates® immunoGrade™ Immunoassay Microplates, Nonsterile, BrandTech®
Supplier: Brandtech
781720 781721 781722 781724 781731 781740 781741 781742 782300 782301 782305 782306
89093-624PK 714.7 CAD
89093-624 89093-626 89093-628 89093-630 89093-632 89093-640 89093-748 89093-750 89093-752 89400-716 89400-718 89400-720 89400-722
BRAND BRANDplates® immunoGrade™ Immunoassay Microplates, Nonsterile, BrandTech®
Microplates Immunology Microplates and Strips
BRANDplates® immunoGrade™ plates are nonsterile plates treated to provide hydrophilic and hydrophobic regions for "high binding" immobiliazation of proteins
  • "High binding" for both hydrophillic and hydrophobic proteins
  • ASNI-SBS formatted
  • 96-well plates embossed for legibility
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. The 96-Well plates have a blue embossed alphanumeric code for quick and easy well identification.
Made of virgin polystyrene,More Product Information