BRAND BRANDplates® pureGrade™ S Microplates, Sterile, BrandTech®
Supplier: Brandtech
781660 781661 781662 781663 781665 781668 781670 781671 781680 781681 781682 781686 781687
89093-608PK 493.99 CAD
89093-608 89093-610 89093-612 89093-614 89093-616 89093-618 89093-620 89093-622 89093-730 89093-732 89093-734 89093-742 89093-744
BRAND BRANDplates® pureGrade™ S Microplates, Sterile, BrandTech®
Microplates HTS Microplates
BRANDplates® pureGrade™ S plates are sterile, untreated plates designed for a variety of standardized applications
  • Untreated, "medium binding" plates
  • ANSI-SLAS formatted
  • Sterility assurance level of 10-6
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. White and black 96-Well plates have a gray embossed alphanumeric code for quick and easy well identification.
Made of virgin polystyrene, these plates are suitable for homogenous assays, screening,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable