BRAND BRANDplates® pureGrade™ Microplates, Non Sterile
Supplier: Brandtech
781600 781601 781602 781603 781604 781605 781607 781608 781610 781611 781620 781621 781622 781626 781627 781640 781641 781642 781614 781615
89093-588PK 438.03 CAD
89093-588 89093-590 89093-592 89093-594 89093-596 89093-598 89093-600 89093-602 89093-604 89093-606 89093-712 89093-714 89093-716 89093-724 89093-726 89093-804 89093-806 89093-808 77674-462 77674-460
BRAND BRANDplates® pureGrade™ Microplates, Non Sterile
Microplates HTS Microplates
BRANDplates® pureGrade™ plates are nonsterile, untreated plates designed for a variety of standardized applications
  • Untreated, "medium binding" plates
  • ANSI-SLAS formatted
  • Non-sterile
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. White and black 96-Well plates feature a gray embossed alphanumeric code for easy well identification.
Made of virgin polystyrene, these plates are suitable for homogenous assays, screening, and storage.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable