Rapid Preparative Chromatography System, Ace Glass Incorporated
Supplier: Ace Glass
5877-60 5877-55 5877-50 5877-65
89053-464EA 1905.9 CAD
89053-464 89053-466 89053-468 89053-470
Rapid Preparative Chromatography System, Ace Glass Incorporated
Chromatography Columns
This rapid preparative chromatography system is suitable for use at pressures up to 50 psig.
This device is available with either a brass or borosilicate glass manifold. The brass manifold has three ⅛" NPT female threads for connection to reservoir, a brass pressure relief valve, and Swagelok tubing connector for connecting to a pressure line. The fourth thread is a ⅛" NPT adapted to ¼" NPT for connecting to a 0 to 60 psig, 2 to ½" Ø pressure gauge.
The glass manifold has two #7 Ace-Threds: one for connecting a pressure line directly using a nylon bushing with PTFE ball for quick disconnect; the other has a nylon-to-PTFE fitting for connecting a brass pressure relief valve. The lower is #15 Ace-Thred for direct connection to the reservoir using a reducer coupling, #15-25 Ace-Thred. The top #15 Ace-Thred on the glass manifold accepts a threaded nylon or PTFE fitting for connection to a pressure gauge.
Available with 25 or 50 mm int.Ø. glass column, 300 mm effective length, with corresponding Ace-Threds. The adapter at bottom of the column is nylon or PTFE with a 'pop-in' polyethylene packing support, 100 µm porosity maximum, and a Luer-Lok fitting takeoff. The reservoir is 2000 or 3000 ml capacity, epoxy coated for safety, pressure tested, with #25 Ace-Threds top and bottom. Connection to the column is via nylon or PTFE coupling. The fitting at top of reservoir is nylon or PTFE, with a ⅛" NPT female thread to accept a Swagelok quick connect-disconnect fitting that in turn connects to the manifold; manifold attaches directly to reservoir using a nylon or PTFE reducer coupling.
Ordering information: CAUTION: Care should be taken when using glass under pressure. Operation behind a shield is highly recommended. Complete system (brass manifold) consists of brass manifold, pressure gauge, brass connector ⅛" NPT to ⅛" ext.Ø tubing, brass connector ⅛" ext.Ø tubing to ¼" NPT female, quick connect-disconnect fitting, pressure relief valve, nylon connector #25 to ⅛" NPT female, epoxy coated reservoir, nylon coupling with FETFE O-rings, column, end fitting with FETFE O-ring and ten feet of ⅛" ext.Ø PTFE tubing. Complete system (glass manifold) consists of glass manifold, pressure gauge, nylon bushing #15 to ¼" NPT female, nylon bushing #7 with PTFE ball, brass connector ⅛" ext.Ø tubing to ¼" NPT female, pressure relief valve, two nylon couplings, with FETFE O-rings, epoxy coated reservoir, column, nylon end fitting and ten feet of ⅛" ext.Ø PTFE tubing. Components are also available separately.