Fitting, Polypropylene, Tee, Compression Union, 1/8" OD; 200/Pk
Supplier: VWR International
VWR Item#MFLX06382-05BLK
Your Organization's Price:
Retrieving
Excellent choice to connect rigid tubing systems.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/8" OD
Compression fittings offer a more permanent installation. Choose the size and material to suit your application needs. <br /><br />Good chemical resistance. Usable from -50 to 250 °F (-46 to 121 °C).
Very good chemical resistance. Rated to 220 psi. Usable from 32 to 212°F (0 to 100°C).