Value Plastics® Fitting, Polypropylene, Straight, Female Luer to Thread Adapter, 1/4-28 UNF; 1000/PK
Supplier: VALUE PLASTICS TE
VWR Item#MFLX45508-66BLK
Your Organization's Price:
Retrieving
Easily transition between luer fittings and UNF or NPT threads.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Female Luer to Threaded, 1/4-28 UNF
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.