Value Plastics® Fitting, Polypropylene, Straight, Female Luer to Smooth Bore Hosebarb, 1/8"; 1000/PK
Supplier: VALUE PLASTICS TE
Easily transition to a luer fitting.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: Female Luer to Smooth Bore Hose Barb, 1/8"