Value Plastics® Fitting, Nylon, Straight, Male Luer Lock to Hose Barb Adapters, 3/32" ID; 1000/PK
Supplier: VALUE PLASTICS TE
Ensure your process integrity with full material traceability.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: Male Luer Lock x 3/32" ID
- Ideal for enhanced fluid path flow
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.