Value Plastics® Fitting, Nylon, Tee, Female Luer x Male Luer; 1000/PK
Supplier: VALUE PLASTICS TE
VWR Item#MFLX45502-85BLK
Your Organization's Price:
Retrieving
Easily transition to a luer fitting.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Female Luer to Female Luer to Male Luer Tee
Easily complete your luer fittings systems with these premium connectors. All female ports may be used with either slip or luer lock connections. Plugs and caps are also available.