Value Plastics® Fitting, Polypropylene, Straight, Female Luer to Hosebarb Adapter, 1/8" ID; 1000/PK
Supplier: VALUE PLASTICS TE
VWR Item#MFLX30800-02BLK
Your Organization's Price:
Retrieving
Easily transition to a luer fitting.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Female luer to 1/8" hose barb with 0.096" diameter orifice