Masterflex® High Flow, 1/2" Barbed Panel Mount, Male, Valved Flow, Polypropylene Housing, Silicone O-Ring, Stainless Spring
Supplier: VWR International
VWR Item#MFLX40255-08
Your Organization's Price:
Retrieving
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a High Flow Female fitting.