Masterflex® High Flow, 3/4" Barbed Panel Mount, Male, Valved Flow, PVDF Housing, Silicone O-Ring, Stainless Spring
Supplier: VWR International
VWR Item#MFLX40255-25
Your Organization's Price:
Retrieving
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a High Flow Female fitting.