Masterflex® High Flow, 3/4" Barbed Panel Mount, Male, Open Flow, Polypropylene Housing
Supplier: VWR International
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a High Flow Female fitting.