Masterflex® Port Plate ½" Barbed Flange in Engage 8402 Cleanroom Manufactured; 10 pack
Supplier: VWR International
VWR Item#MFLX40332-09
Your Organization's Price:
Retrieving
Ribbed base prevents suction blockage and increases rigidity.
  • Barb sizes range from 1/8" to 1"
  • 1-3/4" and 2-1/4" ribbed bases
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.