Masterflex® Console Gear Pump Drive for Micropump A-Mount Pump Head, 5.850 ml/min; 230 VAC
Supplier: Avantor Fluid Handling
VWR Item#MFLX75211-17-IT
Your Organization's Price:
Retrieving
Simple variable speed control handles many fluid handling applications.
  • 100:1 speed-control range controls flow over a wide range
  • Maximum continuous-duty flow rates possible with Micropump® A-mount pump heads
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Remote control of start/stop via DB9 connector on back of drive
  • Maximum flow rate of 5.850 mL/min depending on pump head selected
  • Powered by 230 VAC
Compact, cost-effective drive offers simple variable speed control for many fluid handling applications from general transfer to filtration to chemical injection. Couple with an A-mount gear pump head for pulseless fluid flow.
<b>Ordering information: </b>Pump head is not included.