Masterflex L/S® Easy-Load® Pump Head Replacement Rotor/Bearing Assembly 07514-12 and -32
Supplier: VWR International
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for use with L/S® Standard Pump Heads 07015-10 and -21
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.