Masterflex® Magnetic Drive Centrifugal Pump, 68 GPM/59 ft, Polypropylene; 115/230 VAC
Supplier: VWR International
VWR Item#MFLX75226-32
Your Organization's Price:
Retrieving
Heavy-duty impeller spindle and fluted impeller combine for efficient cooling.
  • Pumps available with or without 56 C-face motors — install the motor you need!
  • Pumps with motor are close-coupled for easy installation and maintenance
  • Ideal for fluid transfer or circulation — use pumps for chemical processing plants and water and wastewater treatment centers
  • Encapsulated magnet
  • Maximum flow rate 68 GPM
  • Maximum feet of head 59 ft
  • Operates on 115/230 VAC
All pumps with motors are set up for hardwiring and are rated for 50/60 Hz duty.
<b>Delivery information: </b>Includes conduit box for hardwiring.