Masterflex® Mechanically Coupled Centrifugal Pump Flange Kit for 1 1/2" and 2" Connections
Supplier: VWR International
Enhance the life of your pump with service flange kits.
- Constructed of stainless steel (SS) parts
- Fits 1 1/2" and 2" connections
Flange kits are competible with your 304 SS mechanically coupled centrifugal pumps.
<b>Delivery information: </b>Includes stainless steel 150# threaded RF companion flanges, gaskets, nuts, bolts, flat washer, and lock washers.