Masterflex® Mechanically Coupled Centrifugal Pump Flange Kit for 1" and 1 1/4" Connections
Supplier: VWR International
VWR Item#MFLX70725-70
Your Organization's Price:
Retrieving
Enhance the life of your pump with service flange kits.
  • Constructed of stainless steel (SS) parts
  • Fits 1" and 1 1/4" connections
Flange kits are competible with your 304 SS mechanically coupled centrifugal pumps.
<b>Delivery information: </b>Includes stainless steel 150# threaded RF companion flanges, gaskets, nuts, bolts, flat washer, and lock washers.